Learn More
ACROBiosystems Inc SARS-CoV-2 Spike NTD Protein, His Tag (B.1.1.529/Omicron) (MALS verified)
Catalog No. 502718907
Encompass
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 502718907 Supplier ACROBiosystems Inc Supplier No. SPDC522D100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.