missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SARS-CoV-2 Proficiency Testing Kit

Catalog No. PE50061KT
Change view
Click to view available options
Content And Storage:
-20°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
PE50061KT -20°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PE50061KT Supplier MilliporeSigma™ Supelco™ Supplier No. PE50061KT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains five samples SARS-CoV-2 sample A through E

  • Sample produced in accordance with ISO/IEC 17043:2010
  • Contains tubes labeled A through E
    • For PCR testing relevant CDC and WHO consensus gene sequence regions
      • Range: 0 to 500,000 copies of RdRp, N, E ang S genes
    • Levels and positive negative assignments of individual tubes are unknown
  • Quality level: 100

Specifications

Certifications/Compliance In accordance with ISO/IEC 17043:2010
Content And Storage -20°C
Format Kit
Type SARS-CoV-2 Proficiency Testing Kit
For Use With (Application) SARS-CoV-2 gene sequencing
Includes Five samples labeled SARS-CoV-2 Sample A through E
Packaging Type Set of 5 tubes each