missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine tert-butyl ester hydrochloride, 97%

CAS: 5616-81-9 | C7H16ClNO2 | 181.66 g/mol

$145.64 - $534.89

Chemical Identifiers

CAS 5616-81-9
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00080936
InChI Key RNLQHMIDSCYLAK-UHFFFAOYSA-N
Synonym h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl
PubChem CID 11252350
IUPAC Name tert-butyl 2-(methylamino)acetate;hydrochloride
SMILES CC(C)(C)OC(=O)CNC.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6317606
View Documents
Thermo Scientific Chemicals
H6317606
5 g
Each for $145.64
Only null left
 
AAH6317614
View Documents
Thermo Scientific Chemicals
H6317614
25 g
Each for $534.89
Only null left
 
Description

Description

Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol (0.3 gram in 2 mL MeOH).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

5616-81-9
181.66
RNLQHMIDSCYLAK-UHFFFAOYSA-N
11252350
CC(C)(C)OC(=O)CNC.Cl
C7H16ClNO2
MFCD00080936
h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl
tert-butyl 2-(methylamino)acetate;hydrochloride

Specifications

5616-81-9
White
CH3NHCH2CO2C(CH3)3·HCl
5 g
Soluble in methanol (0.3 gram in 2 mL MeOH).
CC(C)(C)OC(=O)CNC.Cl
181.66
181.66
Powder
137°C to 141°C
C7H16ClNO2
MFCD00080936
h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl
RNLQHMIDSCYLAK-UHFFFAOYSA-N
tert-butyl 2-(methylamino)acetate;hydrochloride
11252350
97%
Sarcosine tert-butyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only