missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine 98.0+%, TCI America™

Catalog No. p-7132103
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M033225G 25 g
M0332100G 100 g
M0332500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M033225G Supplier TCI America Supplier No. M033225G
Only null left

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym sarcosine, n-methylglycine, sarcosinic acid, methylglycine, glycine, n-methyl, methylaminoacetic acid, n-methylaminoacetic acid, sarcosin, 2-methylamino acetic acid, methylamino acetic acid
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O

Specifications

Melting Point 210°C
Color White
pH 6.0 - 7.0
Quantity 25 g
Formula Weight 89.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sarcosine
EINECSNumber (2)-1205
RTECSNumber VQ2897000
TSCA Yes