missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine methyl ester hydrochloride, 97%

Catalog No. p-7044876
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0857209 10 g
AAL0857218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0857209 Supplier Thermo Scientific Chemicals Supplier No. L0857209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13515-93-0 | C4H10ClNO2 | 139.58 g/mol

It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 13515-93-0
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00038876
InChI Key HQZMRJBVCVYVQA-UHFFFAOYSA-N
Synonym sarcosine methyl ester hydrochloride, methyl 2-methylamino acetate hydrochloride, h-sar-ome.hcl, glycine, n-methyl-, methyl ester, hydrochloride, unii-ypw7yyp04f, ypw7yyp04f, sarcosine methyl ester hcl, methyl n-methylaminoacetate hydrochloride, glycine, n-methyl-, methyl ester, hydrochloride 1:1, sar-ome hcl
PubChem CID 83544
IUPAC Name methyl 2-(methylamino)acetate;hydrochloride
SMILES [H+].[Cl-].CNCC(=O)OC

Specifications

Melting Point ∽105°C (decomposition)
Quantity 10 g
Beilstein 3677128
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 139.58
Percent Purity 98%
Chemical Name or Material Sarcosine methyl ester hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 236-853-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only