missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine methyl ester, hydrochloride, 97%

Catalog No. AC397500500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397500500 50 g Glass bottle
AC397502500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397500500 Supplier Thermo Scientific Chemicals Supplier No. 397500500

CAS: 13515-93-0 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13515-93-0
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00038876
InChI Key HQZMRJBVCVYVQA-UHFFFAOYSA-N
Synonym sarcosine methyl ester hydrochloride, methyl 2-methylamino acetate hydrochloride, h-sar-ome.hcl, glycine, n-methyl-, methyl ester, hydrochloride, unii-ypw7yyp04f, ypw7yyp04f, sarcosine methyl ester hcl, methyl n-methylaminoacetate hydrochloride, glycine, n-methyl-, methyl ester, hydrochloride 1:1, sar-ome hcl
PubChem CID 83544
IUPAC Name methyl 2-(methylamino)acetate;hydrochloride
SMILES [H+].[Cl-].CNCC(=O)OC

Specifications

Melting Point 110.0°C to 125.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 50 g
Formula Weight 139.58
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Sarcosine methyl ester, hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-853-2

RUO – Research Use Only