missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine, 98%

Catalog No. AAA1459436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1459436 500 g
AAA1459422 100 g
AAA145940E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1459436 Supplier Thermo Scientific Chemicals Supplier No. A1459436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol

A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.

Solubility
Soluble in water, methanol. Slightly soluble in ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym sarcosine, n-methylglycine, sarcosinic acid, methylglycine, glycine, n-methyl, methylaminoacetic acid, n-methylaminoacetic acid, sarcosin, 2-methylamino acetic acid, methylamino acetic acid
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O

Specifications

Melting Point ∼208°C (decomposition)
pH 6.1
Odor Odorless
Quantity 500 g
Beilstein 1699442
Sensitivity Hygroscopic
Merck Index 14,8373
Solubility Information Soluble in water,methanol. Slightly soluble in ether.
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material Sarcosine
Show More Show Less
EINECSNumber 203-538-6
RTECSNumber VQ2897000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only