missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Sapphire RT-PCR Breakable 8-Tube Strips with Individual Attached Caps
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling
Specifications
Specifications
| Color | Clear |
| Volume (Metric) | 0.2 mL |
| For Use With (Equipment) | Real-time/quantitative PCR |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Optical (Attached) |
| Product Type | RT-PCR 8-Tube Strips |