Learn More
Azure Biosystems Sapphire FL Standard Optical Module
Description
- Standard Optical Module. Includes laser and emission filter.
Specifications
Specifications
| Wavelength | 375 nm |
| Includes | 452/45 nm Filter |
| Product Type | Optical Module |