missing translation for 'onlineSavingsMsg'
Learn More

Saponin, TCI America™

Catalog No. S001925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001925G Supplier TCI America Supplier No. S001925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8047-15-2
MDL Number MFCD00081981

Specifications

pH 4.80 to 4.90 (50g/L 25°C)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Saponin
RTECSNumber VQ1400000
TSCA No