missing translation for 'onlineSavingsMsg'
Learn More

Saponin, Spray-Dried Powder, Spectrum™ Chemical

Catalog No. 18613078
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613078 50 kg Fiber Drum
18613074 25 g Poly Bottle
18613075 125 g Poly Bottle
18613077 500 g Poly Pail
18613079 2.5 kg Poly Pail
18613076 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613078 Supplier Spectrum Chemical Mfg Cor Supplier No. S564350KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8047-15-2

Specifications

CAS Max % 1
pH 3.8 to 4.6
Packaging Fiber Drum
Quantity 50 kg
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.