missing translation for 'onlineSavingsMsg'
Learn More

Santonin 98.0+%, TCI America™

Catalog No. p-7135918
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S052125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S052125G Supplier TCI America Supplier No. S052125G

Chemical Identifiers

CAS 481-06-1
Molecular Formula C15H18O3
Molecular Weight (g/mol) 246.31
MDL Number MFCD00135865
InChI Key XJHDMGJURBVLLE-BOCCBSBMSA-N
Synonym santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum
PubChem CID 221071
ChEBI CHEBI:16363
IUPAC Name (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione
SMILES C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O

Specifications

Melting Point 173°C
Color White
Quantity 25 g
Formula Weight 246.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Santonin
RTECSNumber LE3150000
TSCA Yes