Learn More
Thermo Scientific Chemicals SANT-1
CAS: 304909-07-7 | C23H27N5, C23H27N5 | 373.50 g/mol
Supplier: Thermo Scientific Chemicals J65294MA
Description
Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling
- High affinity antagonist of smoothened activity
Specifications
| 10 mg | |
| -30°C to -10°C | |
| 304909-07-7 | |
| 373.49g/mol | |
| Moisture and Light Sensitive | |
| Signal Transduction Reagent | |
| FOORCIAZMIWALX-UHFFFAOYSA-N | |
| N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine |
| Pale Yellow | |
| Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity | |
| C23H27N5 | |
| (4-Benzyl-piperazin-1-yl)-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-ylmethylene)-amine | |
| Soluble in DMSO with warming | |
| Solid | |
| CC1=NN(C(C)=C1C=NN1CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1 | |
| 373.50 |
Chemical Identifiers
| 304909-07-7 | |
| 373.50 | |
| (4-Benzyl-piperazin-1-yl)-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-ylmethylene)-amine | |
| CC1=NN(C(C)=C1C=NN1CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1 |
| C23H27N5 | |
| FOORCIAZMIWALX-UHFFFAOYSA-N | |
| N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only