missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sanicult™ Environmental Transport Kits

Catalog No. p-4525682
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
R723140 10 mL
R723141 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R723140 Supplier Thermo Scientific™ Supplier No. RE723140
Only null left

Thermo Scientific™ Remel Sanicult Environmental Transport Kits can be used to sample most surfaces or substances.

Thermo Scientific™ Remel Sanicult Environmental Transport Kits can be used to sample most surfaces or substances.

  • Designed for hard-to-reach surfaces, viscous liquids, semi-solids, and powders
  • Readily transported with room temperature storage
  • Manufacturer: Starplex

Ordering Alerts:

Supplied one swab per envelope; 100 envelopes/pack.

Includes:

Screw cap sampling vial with sterile diluent; Sterile swab in a screw cap tube.

Specifications

Format Swab
For Use With (Application) Healthcare
Quantity 100 Pk.
Storage Requirements Stable for up to one year from the date of manufacture when stored at room temperature.
Volume (Metric) 10 mL