missing translation for 'onlineSavingsMsg'
Learn More

Sand, White Quartz, Honeywell Fluka™

Catalog No. 6002246
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002246 500 g
6002245 1 kg
6002247 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6002246 Supplier Honeywell Chemicals Supplier No. 274739500G
Only null left

50-70 mesh particle size

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White
Quantity 500 g
Linear Formula SiO2
UN Number NONH for all modes of transport
IUPAC Name dioxosilane
Formula Weight 60.08g/mol
Chemical Name or Material Sand, White Quartz
EINECSNumber 238-878-4
RTECSNumber VV7330000