missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, White Quartz, Honeywell Fluka™
50-70Â mesh particle size
Supplier: Honeywell-Fluka 2747391KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sand, White Quartz | |
| 14808-60-7 | |
| O2Si | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 |
| White | |
| 1 kg | |
| SiO2 | |
| NONH for all modes of transport | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| dioxosilane | |
| 24261 | |
| 60.08g/mol |
Chemical Identifiers
| 14808-60-7 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| 24261 | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| CHEBI:30563 |
Safety and Handling
EINECSNumber : 238-878-4
RTECSNumber : VV7330000