missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Sea (Washed), Fisher Chemical™
Specifications
Specifications
| Boiling Point | 2230°C |
| Melting Point | 1610°C |
| Color | Beige |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Density | 2.65g/cm³ |
| Chemical Name or Material | Sand, Sea, Washed |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting. NFPA Health:1 Flammability:0 Instability:0 |