missing translation for 'onlineSavingsMsg'
Learn More

Sand, Reagents

Catalog No. 01337078
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337078 3 kg
01337077 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337078 Supplier Labchem Inc Supplier No. C22990003A5

missing translation for 'validMainframeMsg'

Sand, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Boiling Range 2230°C
Melting Point 1610°C
Color White to Light Brown
Boiling Point 2230°C
Melting Range 1610°C
Packaging Poly Bottle
Quantity 3 kg
Specific Gravity 2.4
Grade Technical
Physical Form Solid
Chemical Name or Material Sand
Show More Show Less