Learn More
Thermo Scientific™ Sampling Manifold Accessory for HyPerforma™ Single-Use Bioreactors, with Luer locks
Catalog No. SH3126302
Change view
Click to view available options
Capacity (Metric):
50/100 mL
Quantity:
1 Ea.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
SH3126302 50/100 mL 10/Pk.
SH3126301 50/100 mL 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3126302 Supplier Thermo Scientific™ Supplier No. SH31263.02
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
Specifications
Capacity (Metric) 50/100 mL
Quantity 10/Pk.
Tubing Style C-Flex