Learn More
Chemglass Inc Vial, Sample, Pre-Cleaned, 40mL, Amber, 28x95mm, GPI 24-400, with PTFE Lined Screw Cap
Supplier: Chemglass Inc CG490705
Vial, Sample, Pre-Cleaned, 40mL, Amber, 28x95mm, GPI 24-400, with PTFE Lined Screw Cap. Clear and amber sample vials are pre-cleaned and certified for volatile organic analysis. Vials are fully assembled with green polypropylene closure and 0.125 PTFE silicone septa. Made from Type 1 borosilicate glass. Certificate of analysis is included in each case. Packed 72 pieces per case.