Learn More
Chemglass Inc 20mL Scintillation Vial, 28mm OD x 57mm Height, 24-400 Thread Size
Supplier: Chemglass Inc CG490004
20mL Scintillation Vial, 28mm OD x 57mm Height, 24-400 Thread Size. Sample vials packaged in convenient LAB-PACS of 144. Made from Type 1 Borosilicate glass. Full height partitions minimize breakage. Solid PTFE Lined Caps.