Learn More
Perkin Elmer Canada PMMA Sample Test Kit - Qty. 5
Catalog No. 502016538
Encompass
The DMA analyzers require the following calibration test samples to ensure optimal performance: PMMA, Steel, PVC, HDPE, Polycarb, PET Film, and G10 Epoxy.
Catalog No. 502016538 Supplier Perkin Elmer Canada Supplier No. N5330316
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The DMA analyzers require the following calibration test samples to ensure optimal performance: PMMA, Steel, PVC, HDPE, Polycarb, PET Film, and G10 Epoxy.