Learn More
Chemplex Industries SAMPLE CUPS 40.1MM DIA. DBL OP DFS Item
Supplier:  Chemplex Industries CMP2143
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 40.1 Dia., mm, Double open ended, 41.1mm Max O.D., Internal Overflow Reservoir
Catalog No. 50249415
Only null left
Explore available promotions