Learn More
Waters Limited 96-well Sample Collection Plate, 2 mL, 50/pk
Catalog No. 50785854
Encompass
96-well sample collection plate with 2 mL well. This plate is SBS/ANSI compliant for robot compatible systems. This plate can also serve as a collection plate for 96-well SPE and filtration-plate formats.
Catalog No. 50785854 Supplier Waters Limited Supplier No. WAT058958
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
96-well sample collection plate with 2 mL well. This plate is SBS/ANSI compliant for robot compatible systems. This plate can also serve as a collection plate for 96-well SPE and filtration-plate formats.