Learn More
Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers

Prevent bio-hazardous spills with these leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Supplier: Thermo Scientific™ 02 1105
Description
Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- 90mL (3 oz.); Opening: 53mm
- Dual-threaded caps create improved sealing
- Sterilized by gamma irradiation, which eliminates potential contamination
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Stackable containers are easy to store, saving lab space
- Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Chemical Identifiers
| Yes | |
| Cylindrical | |
| 53 mm | |
| 53 mm |
| Virgin Polypropylene | |
| 90 mL | |
| Bio-Tite Lid |
Specifications
| Specimen Container | |
| Virgin Polypropylene | |
| Non-sterile | |
| Yellow | |
| Bio-Tite Lid | |
| 53 mm | |
| 300/Cs., Pre-assembled |
| Yes | |
| Cylindrical | |
| 90 mL | |
| 53 mm | |
| Cap and container assembled | |
| Non-tabbed |