Learn More
Thermo Scientific™ Samco™ Transfer Pipettes
Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
Supplier:  Thermo Scientific™ 2041S
Catalog No. 22-610-501
Only null left
Explore available promotions
Description
Description
Prevent cross-contamination with Thermo Scientific™ Samco™ Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface reduces loss of cells and valuable proteins due to binding. They can be frozen in liquid nitrogen, or heat sealed for storage.
Recommended for:
Tissue culture and applications, routing sampling of serum and plasma, blood culture, urine analysis
Product Suggestions
Product Suggestions