Learn More
Thermo Scientific™ Samco™ SW-3000 24-hour Urine Collection Containers
Description
Alleviate mess and exposure when handling urine collection and sampling with the Thermo Scientific™ Samco™ 24-hour Urine Collection product line. Thermo Scientific Samco SW-3000 containers are amber tinted to protect from light exposure and are inert to normal preservatives.
- Large handle for easy transportation
- Custom cap and neck design to prevent leaking
- Accurate scale for proper measurement, eliminating the need to transfer samples into a graduated cylinder
Specifications
Specifications
| Product Type | Urine Collection Container |
| Closure Type | Wide-mouth Screw Cap with Integral Spout |
| Graduated | Yes |
| Color | Amber |
| Description | 24-hour Urine Container |
| Includes | Container Only |
| No. per Pack | 40 Pk. |
| Volume (Metric) | 3000 mL |
| Green Features | Less Waste |