missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Special Purpose Slurry Pipette
GREENER_CHOICE

Catalog No. 14754525
Catalog No. 14754525 Supplier Thermo Scientific™ Supplier No. MB993

Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles.

Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles. Single-unit design features a built-in pipettes bulb that eliminates need for plugging, packaging, and autoclaving; prevents cross-contamination.

Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles. Single-unit design features a built-in pipettes bulb that eliminates need for plugging, packaging, autoclaving; prevents cross-contamination.

Specifications

Bulb Draw (Metric) 3.3 mL
Description Special Purpose Slurry Pipette; 7.5mL Pipettes
Diameter (Metric) Outer 5.08 mm
Sterility Non-sterile
Drops/mL 13
Quantity 450/Pk., 10 Packs/Cs., 4500/Cs.
Volume (Metric) 7.5 mL
Length (Metric) 13.23 cm