missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Samco™ Narrow Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers
Prevent bio-hazardous spills and conserve storage space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.
$451.09 - $451.09
Specifications
| Product Type | Specimen Container |
|---|---|
| Closure Included | Yes |
| Material | Virgin Polypropylene |
| Capacity (Metric) | 60 mL |
| Closure Size | 48 mm |
| Catalog Number | Mfr. No. | Sterility | Closure Color | Label | No. per Pack | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Closure Color | Label | No. per Pack | Price | Quantity | ||
|
13-711-63
|
Thermo Scientific™
030193 |
Non-sterile | Yellow | No Label | 100/Pk., Pre-assembled |
Case of 500 Each for $451.09
|
|
||
Description
Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:- Dual-threaded caps create leak-resistant seal for superior safety and handling
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Virgin polypropylene allows higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Available Options:
- Nonsterile or gamma irradiation sterilized
- Labled or non-labled
- Bulk or individually packed
Specifications
| Specimen Container | |
| Virgin Polypropylene | |
| 48 mm |
| Yes | |
| 60 mL | |
| 48 mm |