Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Supplier: Thermo Scientific™ 966NL
Description
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
- Shatterproof, nontoxic polyethylene
- Nonsterile
- Low affinity surface
- Freezer ready
Specifications
| 300μL Exact Volume Transfer Pipette | |
| LDPE | |
| No | |
| 300 μL | |
| Exact Volume Transfer Pipet |
| 3.6 mm | |
| Non-sterile | |
| Bulk 500/Pk 10Pk/Cs 5000/Cs | |
| 9.5 cm | |
| Single-use |