Learn More
Samarium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-19-9 | Sm | 150.36 g/mol
Supplier: Thermo Scientific Chemicals 040297DH
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
| Samarium rod | |
| 7440-19-9 | |
| 25 mm | |
| Sm | |
| samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
| [Sm] | |
| 150.36 | |
| CHEBI:33374 | |
| Odorless |
| 6.35mm (0.25 in.) dia. | |
| Cast Rod | |
| (metals basis excluding Ta) | |
| MFCD00011233 MFCD00151299 | |
| KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
| samarium | |
| 23951 | |
| 99.9% | |
| Mylar under Argon |
Chemical Identifiers
| 7440-19-9 | |
| 150.36 | |
| KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
| 23951 | |
| samarium |
| Sm | |
| MFCD00011233 MFCD00151299 | |
| samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
| CHEBI:33374 | |
| [Sm] |
Safety and Handling
P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
H228-H260
DOTInformation : Hazard Class: 4.3; Packaging Group: III
EINECSNumber : 231-128-7
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only