missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Catalog No. p-7051148
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4056709 10 g
AA4056718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4056709 Supplier Thermo Scientific Chemicals Supplier No. 04056709
Only null left

CAS: 13569-60-3 | Cl3H2O13Sm | 466.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13569-60-3
Molecular Formula Cl3H2O13Sm
Molecular Weight (g/mol) 466.71
MDL Number MFCD00049769
InChI Key ONSMNAINZIBBGR-UHFFFAOYSA-K
Synonym samarium 3+ ;triperchlorate, samarium iii perchlorate solution, trihyperchloric acid samarium iii salt, samarium iii perchlorate w/w aqueous solution, reagent grade
IUPAC Name samarium(3+) hydrate triperchlorate
SMILES O.[Sm+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Formula Weight 556.80 (448.70 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Samarium(III) perchlorate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 236-986-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only