missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Catalog No. AA4066506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4066506 5 g
AA4066514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4066506 Supplier Thermo Scientific Chemicals Supplier No. 04066506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64438-53-5 | C15H21O6Sm | 447.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64438-53-5
Molecular Formula C15H21O6Sm
Molecular Weight (g/mol) 447.69
MDL Number MFCD00792904
InChI Key JTZJMDRBZZVNSP-UHFFFAOYSA-N
Synonym samarium 2,4-pentanedionate, samarium iii acetylacetonate hydrate, samarium iii 2,4-pentanedionate hydrate, samarium iii acetylacetonate hydrate trace metals basis, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy samario oxy pent-3-en-2-one hydrate
PubChem CID 16213768
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;samarium;hydrate
SMILES [Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 447.68 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium(III) 2,4-pentanedionate hydrate
EINECSNumber 238-632-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only