missing translation for 'onlineSavingsMsg'
Learn More

Samarium(II) Iodide (ca. 0.1mol/L in Tetrahydrofuran), TCI America™

Catalog No. S049425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S049425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S049425ML Supplier TCI America Supplier No. S049425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32248-43-4
Molecular Formula I2Sm

Specifications

Color Blue
Physical Form Liquid
Quantity 25 mL
UN Number 2056
Formula Weight 404.17
Chemical Name or Material Samarium(II) Iodide (ca. 0.1mol/L in Tetrahydrofuran)