missing translation for 'onlineSavingsMsg'
Learn More

Samarium(II) iodide, 0.07 - 0.12M in THF, stab.

CAS: 32248-43-4

$187.12 - $686.70

Chemical Identifiers

CAS 32248-43-4
MDL Number MFCD00058873
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA22360AE
View Documents
Thermo Scientific Chemicals
022360AE
100 mL
Each for $187.12
Only null left
 
AA22360AP
View Documents
Thermo Scientific Chemicals
022360AP
500 mL
Each for $686.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32248-43-4
MFCD00058873

Specifications

32248-43-4
Ether-like
100 mL
Air and moisture sensitive
0.07 to 0.12M in THF
Samarium(II) iodide, Stabilized
0.922 g/mL
MFCD00058873
UN3271
404.21
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H260-H314-H318
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302-H319-H335-H336-H351-H373

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 203-726-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only