missing translation for 'onlineSavingsMsg'
Learn More

Samarium foil, 0.75mm (0.03 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. AA43218FF
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43218FF 25 x 25 mm
AA43218FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43218FF Supplier Thermo Scientific Chemicals Supplier No. 043218FF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-19-9 | Molecular Formula: Sm | Molecular Weight: 150.36 g/mol

Used in control rods in nuclear reactors, as a catalyst in organic chemistry, a pyrophoric metal in lighter flints, and a core in carbon arc-lamp electrodes. Also used in alloys with cobalt to make strong permanent magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈3.7g/25x25mm
Form Foil
Solubility Information Insoluble in water.
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium foil, 0.75mm (0.03 in.) thick
Hazard Category H228-H260
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: III
EINECSNumber 231-128-7
TSCA Yes
storageNote1
  • Packaged in mylar under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only