missing translation for 'onlineSavingsMsg'
Learn More

3B Scientific™ SAM4 Auscultation Manikin Dark Skin
GREENER_CHOICE

Catalog No. S012543
Change view
Click to view available options
Dark Skin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S012543 Dark Skin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012543 Supplier 3B Scientific™ Supplier No. 1025087

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The latest advancement in auscultation training from Cardionics.

  • Introducing SAM4, the latest advancement in auscultation training from Cardionics.
  • SAM4 builds upon the success of its predecessors, including SAM Basic, SAM II, and SAM 3G, and brings exclusive new features to elevate the auscultation learning experience.

Specifications

Product Type Auscultation Simulator
For Use With (Application) Teaching and Learning Auscultation
Color Dark Skin