Learn More
Bullard™ SALUS HC Power Source Replacement Parts and Accessories
Description
- Power source replacement parts for the SALUS HC powered air-purifying respirator
- High-capacity battery includes battery level indicator
- Single battery charger and power supply
- Replacement battery housing lid keeps the battery secure in the unit
Specifications
Specifications
| Battery Type | Lithium-ion |
| Description | Replacement Parts |
| Product Type | Battery Pack |
| Model | SALHCBAT |