Learn More
Bullard™ SALUS HC Power Source Replacement Parts and Accessories
Description
- Power source replacement parts for the SALUS HC powered air-purifying respirator
- High-capacity battery includes battery level indicator
- Single battery charger and power supply
- Replacement battery housing lid keeps the battery secure in the unit
Specifications
Specifications
| Description | Replacement Parts |
| Product Type | Battery Charger |
| For Use With (Equipment) | SALUS HC System |
| Includes | Single Battery Charger |
| Model | SALHCBC1 |