Learn More
Bullard™ SALUS HC Power Source Replacement Parts and Accessories
Catalog No. 19087969
Change view
Click to view available options
Product Type:
Battery Charger
Battery Pack
Model:
SALHCBAT
SALHCBC1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Model
19087969 Battery Pack SALHCBAT
19087971 Battery Charger SALHCBC1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19087969 Supplier Bullard™ Supplier No. SALHCBAT
Only null left
Choose from a variety of power sources offered for use with the SALUS HC powered air-purifying respirator.
  • Power source replacement parts for the SALUS HC powered air-purifying respirator
  • High-capacity battery includes battery level indicator
  • Single battery charger and power supply
  • Replacement battery housing lid keeps the battery secure in the unit
Specifications
Battery Type Lithium-ion
Description Replacement Parts
Product Type Battery Pack
Model SALHCBAT