missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ SALUS HC Carriage Release Hook, White, Pack of 2

Catalog No. 19087973
Change view
Click to view available options
For use with: SALUS HC System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19087973 For use with: SALUS HC System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19087973 Supplier Bullard™ Supplier No. SALHCHK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pack of 2 SALUS HC Carriage Release Hooks.

  • White
  • Pack of 2

Specifications

Color White
Disposable Reusable
Product Type Carriage Release Hook
Model SALHCHK
Quantity 2/Pk.