missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Salubrinal

Catalog No. p-7082396
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64192LB0 5 mg
AAJ64192MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64192LB0 Supplier Thermo Scientific Chemicals Supplier No. J64192LB0
Only null left

Salubrinal is a selective Inhibitor of eIF2α (eukaryotic translation initiation factor 2 α-subunit) dephosphorylation, inhibiting global protein translation. Salubrinal is a specific inhibitor of ER Stress induced apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 405060-95-9
Molecular Formula C21H17Cl3N4OS
Synonym eIF-2alpha Inhibitor; SAL

Specifications

Quantity 5 mg
Description Pale Brown
Content And Storage 2°C to 8°C
Formula Weight 479.81g/mol
Sensitivity Light Sensitive
Product Type Apoptosis Reagent
Form Powder
InChI Key LCOIAYJMPKXARU-UHFFFAOYNA-N
SMILES ClC(Cl)(Cl)C(NC(=S)NC1=C2N=CC=CC2=CC=C1)NC(=O)C=CC1=CC=CC=C1
IUPAC Name 3-phenyl-N-(2,2,2-trichloro-1-{[(quinolin-8-yl)carbamothioyl]amino}ethyl)prop-2-enamide
Molecular Weight (g/mol) 479.80
Show More Show Less
Recommended Storage Keep cold

RUO – Research Use Only