Learn More
New England Biolab Ltd Salt-T4 DNA Ligase 100000 units
Catalog No. 502024036
Encompass
A salt tolerant variant of T4 DNA Ligase, Salt-T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA and is designed to function at higher salt concentrations than wild type T4 DNA Ligase. This enzyme will join cohesive end termini at salt concentrations as high as 300 mM without any loss in activity. This enzyme is insensitive to salt carried over from other reaction components (vector or insert DNA) and allows ligation reactions to proceed in alternative reaction buffers with higher levels of salt (e.g., NEBuffer r3.1).
Catalog No. 502024036 Supplier New England Biolab Ltd Supplier No. M0467L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A salt tolerant variant of T4 DNA Ligase, Salt-T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA and is designed to function at higher salt concentrations than wild type T4 DNA Ligase. This enzyme will join cohesive end termini at salt concentrations as high as 300 mM without any loss in activity. This enzyme is insensitive to salt carried over from other reaction components (vector or insert DNA) and allows ligation reactions to proceed in alternative reaction buffers with higher levels of salt (e.g., NEBuffer r3.1).