missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Salmeterol xinafoate
Chemical Identifiers
| CAS | 94749-08-3 |
|---|---|
| Molecular Formula | C36H45NO7 |
| Molecular Weight (g/mol) | 603.76 |
| MDL Number | MFCD00897708 |
| InChI Key | XTZNCVSCVHTPAI-UHFFFAOYNA-N |
| Synonym | salmeterol xinafoate, salmetedur, arial, serevent diskus, asmerole, betamican, dilamax, inaspir, ultrabeta, beglan |
| PubChem CID | 56801 |
| IUPAC Name | 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid |
| SMILES | OC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC462171000
|
Thermo Scientific™
462171000 |
100mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC462175000
|
Thermo Scientific™
462175000 |
500mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 94749-08-3 | |
| 603.76 | |
| XTZNCVSCVHTPAI-UHFFFAOYNA-N | |
| 56801 | |
| OC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1 |
| C36H45NO7 | |
| MFCD00897708 | |
| salmeterol xinafoate, salmetedur, arial, serevent diskus, asmerole, betamican, dilamax, inaspir, ultrabeta, beglan | |
| 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid |
Specifications
| 94749-08-3 | |
| MFCD00897708 | |
| salmeterol xinafoate, salmetedur, arial, serevent diskus, asmerole, betamican, dilamax, inaspir, ultrabeta, beglan | |
| XTZNCVSCVHTPAI-UHFFFAOYNA-N | |
| 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid | |
| 56801 | |
| Salmeterol xinafoate |
| C36H45NO7 | |
| 100mg | |
| Solubility in water: soluble. Other solubilities: soluble in dmso, methanol | |
| OC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1 | |
| 603.76 | |
| 603.75 |
RUO â Research Use Only