missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Saline 0.45% Solution Tubes
Reduce sample preparation time for automated identification and susceptibility systems using convenient, pre-filled and ready-to-use 0.45% saline tubes.
$939.34 - $1082.69
Specifications
| Color | Clear liquid colorless |
|---|---|
| Content And Storage | 2 to 25°C |
| Format | Tube |
| Manufacturing Origin | United States |
| Product Type | Buffer Solution |
| Catalog Number | Mfr. No. | Quantity | Format | Manufacturing Origin | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Format | Manufacturing Origin | Price | Quantity | |||
|
R064462
|
Thermo Scientific™
RE064462 |
100/Pk. | Tube | United States |
Case of 100 Each for $1,082.69
|
|
|||
|
R064464
|
Thermo Scientific™
RE064464 |
100/Pk. | Tube | United States |
Case of 100 Each for $939.34
|
|
|||
Description
Save time when preparing your samples for automated identification and susceptibility systems using Thermo Scientific™ Saline 0.45% Solution Tubes.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes.- Easily adjust inoculum density with a clear tube
- Enhance sample security while promoting safe handling with a reliable screw cap on each tube
- Achieve versatility and flexibility with fill volumes in 1.8mL and 3.0mL
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
| Clear liquid colorless | |
| Tube | |
| Buffer Solution |
| 2 to 25°C | |
| United States | |
| 100/Pk. |