missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Saline 0.45% Solution Tubes

Catalog No. R064462
Change view
Click to view available options
Quantity:
100/Pk.
Format:
Tube
Manufacturing Origin:
United States
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Manufacturing Origin
R064462 100/Pk. Tube United States
R064464 100/Pk. Tube United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R064462 Supplier Thermo Scientific™ Supplier No. RE064462

Reduce sample preparation time for automated identification and susceptibility systems using convenient, pre-filled and ready-to-use 0.45% saline tubes.

Save time when preparing your samples for automated identification and susceptibility systems using Thermo Scientific™ Saline 0.45% Solution Tubes.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes.
  • Easily adjust inoculum density with a clear tube
  • Enhance sample security while promoting safe handling with a reliable screw cap on each tube
  • Achieve versatility and flexibility with fill volumes in 1.8mL and 3.0mL

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Color Clear liquid colorless
Content And Storage 2 to 25°C
Format Tube
Manufacturing Origin United States
Product Type Buffer Solution
Fill Volume 1.8 mL
Packaging Type Glass tube with screw cap
Quantity 100/Pk.