missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid, ACS Reagent (2-Hydroxybenzoic Acid), Reagents

Catalog No. p-200021134
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338163 100 g
01338161 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338163 Supplier Labchem Inc Supplier No. C1113000100C2
Only null left

Salicylic Acid, ACS Reagent (2-Hydroxybenzoic Acid), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym 2-Hydroxybenzoic Acid, 2-Carboxyphenol, o-Hydroxybenzoic Acid
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 159°C
Boiling Point 211°C
Color Colorless/White
Grade ACS
Quantity 100 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 1.443
Chemical Name or Material Salicylic Acid, ACS Reagent (2-Hydroxybenzoic Acid)