missing translation for 'onlineSavingsMsg'
Learn More

Salicylic acid, 99+%, ACS reagent, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 419220020

Catalog No. AC419220020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Specifications

Salicylic acid
99+%
MFCD00002439
15, 8469
YGSDEFSMJLZEOE-UHFFFAOYSA-N
2-hydroxybenzoic acid
338
138.12
ACS Reagent
157°C
Plastic Bottle
69-72-7
C7H6O3
10, 43
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
OC(=O)C1=CC=CC=C1O
138.12
CHEBI:16914
≥99%
2kg
Authentic
0.01% max.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Causes serious eye damage.

Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0

SIGNAL WORD: DANGER!

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only