missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde 98.0+%, TCI America™

Catalog No. p-7135857
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S027525G 25 g
S0275500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S027525G Supplier TCI America Supplier No. S027525G
Only null left

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point 2°C
Boiling Point 196°C
Quantity 25 g
Formula Weight 122.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
EINECSNumber (3)-1183&(3)-2660
RTECSNumber VN5250000
TSCA Yes