missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylaldehyde 98.0+%, TCI America™
Supplier: TCI America S0004100G
Specifications
| Salicylaldehyde | |
| 2°C | |
| 196°C | |
| MFCD00003317 | |
| salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy | |
| OC1=CC=CC=C1C=O | |
| 122.12 | |
| CHEBI:16008 | |
| ≥98.0% (GC) |
| 90-02-8 | |
| Yellow | |
| C7H6O2 | |
| 100 g | |
| SMQUZDBALVYZAC-UHFFFAOYSA-N | |
| 2-hydroxybenzaldehyde | |
| 6998 | |
| 122.12 | |
| Liquid |
Chemical Identifiers
| 90-02-8 | |
| 122.12 | |
| SMQUZDBALVYZAC-UHFFFAOYSA-N | |
| 6998 | |
| 2-hydroxybenzaldehyde |
| C7H6O2 | |
| MFCD00003317 | |
| salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy | |
| CHEBI:16008 | |
| OC1=CC=CC=C1C=O |
Safety and Handling
EINECSNumber : (3)-1183&(3)-2660
RTECSNumber : VN5250000
TSCA : Yes